Thermo Scientific Chemicals

Starch, for biochemistry, potato, hydrolyzed for electrophoresis

Catalog number: 215121000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Starch, for biochemistry, potato, hydrolyzed for electrophoresis

Catalog number: 215121000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 215121000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-25-8
IUPAC Name
2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-UHFFFAOYNA-N
SMILES
OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Appearance (Form)
Crystalline powder
Loss on drying
=<20 %
Residue after ignition
=<1 %
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text