Thermo Scientific Chemicals

Prussian Blue, pure

Catalog number: 215211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Prussian Blue, pure

Catalog number: 215211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215211000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14038-43-8
IUPAC Name
tetrairon(3+) tris(λ²-iron(2+)) octadecakis(iminomethanide)
Molecular Formula
C18Fe7N18
InChI Key
DCYOBGZUOMKFPA-UHFFFAOYSA-N
SMILES
[Fe++].[Fe++].[Fe++].[Fe+3].[Fe+3].[Fe+3].[Fe+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Appearance (Color)
Dark blue
UV spectrum
Complies
Infrared spectrum
Conforms
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text