Thermo Scientific Chemicals

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Catalog number: 215271000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Catalog number: 215271000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 215271000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
111-31-9
IUPAC Name
hexane-1-thiol
Molecular Formula
C6H14S
InChI Key
PMBXCGGQNSVESQ-UHFFFAOYSA-N
SMILES
CCCCCCS
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Density
(20°C) 0.836 to 0.844 g/ml
Appearance (Form)
Liquid
Refractive index
1.4480 to 1.4520 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text