Thermo Scientific Chemicals

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%, Thermo Scientific Chemicals

Catalog number: 215280050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%, Thermo Scientific Chemicals

Catalog number: 215280050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 215280050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69304-37-6
IUPAC Name
chloro({[chlorobis(propan-2-yl)silyl]oxy})bis(propan-2-yl)silane
Molecular Formula
C12H28Cl2OSi2
InChI Key
DDYAZDRFUVZBMM-UHFFFAOYSA-N
SMILES
CC(C)[Si](Cl)(O[Si](Cl)(C(C)C)C(C)C)C(C)C
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=95 %
Refractive index
1.4525 to 1.4575 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text