L(+)-2-Aminobutyric acid, 98%, Thermo Scientific Chemicals
L(+)-2-Aminobutyric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L(+)-2-Aminobutyric acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 215380050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1492-24-6
IUPAC Name2-aminobutanoic acid
Molecular FormulaC4H9NO2
InChI KeyQWCKQJZIFLGMSD-UHFFFAOYNA-N
SMILESCCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
Sulfated ash=<0.1 %
Water=<0.5 % (K.F.)
Specific optical rotation+20.2° to +23° (20°C, 589 nm) (c=2, 5 N HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only