Thermo Scientific Chemicals

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, Thermo Scientific Chemicals

Catalog number: 215410010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, Thermo Scientific Chemicals

Catalog number: 215410010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 215410010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
8001-54-5
Molecular Formula
C9H13ClN
SMILES
[Cl-].C[N+](C)(*)CC1=CC=CC=C1
Molecular Weight (g/mol)
170.66
Appearance (Color)
Colorless to white to yellow
Sulfated ash
=<0.5 %
Appearance
May contain yellowish-white fragments
Assay
95.0 to 104.0 % (on anhydrous basis)
Appearance (Form)
Solid or thick gel

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text