Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
Thermo Scientific Chemicals

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33

CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 215411000
Price (USD)
42.65
Online Exclusive
47.10
Save 4.45 (9%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.65
Online Exclusive
47.10
Save 4.45 (9%)
Each
Add to cart
Chemical Identifiers
CAS8001-54-5
Molecular FormulaC9H13ClN
SMILES[Cl-].C[N+](C)(*)CC1=CC=CC=C1
Molecular Weight (g/mol)170.66
SpecificationsSpecification SheetSpecification Sheet
Total amines=<3.6 %
Appearance (Color)Colorless to white to yellow
Sulfated ash=<0.5 %
AppearanceMay contain yellowish-white fragments
Assay95.0 to 104.0 % (on anhydrous basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link