Clear colorless to yellow to orange to light brown
Refractive index
1.5440 to 1.5490 (20°C, 589 nm)
Stabilizer
Copper
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.