1x1 image pixel for data collection
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C) | CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 215470010
Price (USD)
239.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
239.00
Each
Add to cart
Chemical Identifiers
CAS7758-11-4
IUPAC Namedipotassium hydrogen phosphate
Molecular FormulaHK2O4P
InChI KeyZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 %
Heavy metals (as Pb)=<5 ppm
Appearance (Form)Crystalline powder
pH8.5 to 9.6 (5 % at 20°C)
Loss on drying=<1 % (130°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link