Thermo Scientific Chemicals

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 215470050
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 215470050
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 215470050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7758-11-4
IUPAC Name
dipotassium hydrogen phosphate
Molecular Formula
HK2O4P
InChI Key
ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES
[K+].[K+].OP([O-])([O-])=O
Loss on drying
=<1 % (130°C)
Chloride (Cl)
=<50 ppm
Iron (Fe)
=<50 ppm
Appearance (Form)
Crystalline powder
pH
8.5 to 9.6 (5 % at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text