Thermo Scientific Chemicals

Phenyl chlorothionocarbonate, 99%, Thermo Scientific Chemicals

Catalog number: 215490250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenyl chlorothionocarbonate, 99%, Thermo Scientific Chemicals

Catalog number: 215490250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215490250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1005-56-7
IUPAC Name
phenyl chloromethanethioate
Molecular Formula
C7H5ClOS
InChI Key
KOSYAAIZOGNATQ-UHFFFAOYSA-N
SMILES
ClC(=S)OC1=CC=CC=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.5800 to 1.5830 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text