Sodium fumarate, 98%, Thermo Scientific Chemicals
Sodium fumarate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fumarate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 215535000
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
Add to cart
Sodium fumarate, 98%, Thermo Scientific Chemicals
Catalog number215535000
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium (2Z)-but-2-enedioate
Molecular FormulaC4H2Na2O4
InChI KeyMSJMDZAOKORVFC-UAIGNFCESA-L
SMILES[Na+].[Na+].[O-]C(=O)\C=C/C([O-])=O
Molecular Weight (g/mol)160.04
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only