Thermo Scientific Chemicals

Tetracosanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 215640050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetracosanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 215640050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
557-59-5
IUPAC Name
tetracosanoic acid
Molecular Formula
C24H48O2
InChI Key
QZZGJDVWLFXDLK-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCC(O)=O
Infrared spectrum
Conforms
Appearance (Color)
White
GC
>=98.5 %
Appearance (Form)
Powder or crystals or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text