Thermo Scientific Chemicals

Tripalmitin, 99%, Thermo Scientific Chemicals

Catalog number: 215670010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Tripalmitin, 99%, Thermo Scientific Chemicals

Catalog number: 215670010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215670010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
555-44-2
IUPAC Name
1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular Formula
C51H98O6
InChI Key
PVNIQBQSYATKKL-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
Appearance (Color)
White to almost white
Infrared spectrum
Conforms
HPLC
>=98.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text