1x1 image pixel for data collection
Aluminum oxide, 99.7+%, extra pure
Aluminum oxide, 99.7+%, extra pure
Aluminum oxide, 99.7+%, extra pure
Aluminum oxide, 99.7+%, extra pure
Thermo Scientific Chemicals

Aluminum oxide, 99.7+%, extra pure

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 215700010
Price (USD)
69.70
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
69.70
Each
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sodium oxide (Na2O)=<0.1 %
Appearance (Color)White
Aluminium oxide (Al2O3)>=99.70 %
Iron(III)oxide (Fe2O3)=<0.03 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection