Aluminum oxide, 99.7+%, extra pure, Thermo Scientific Chemicals
Aluminum oxide, 99.7+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, 99.7+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 215700050
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
Add to cart
Aluminum oxide, 99.7+%, extra pure, Thermo Scientific Chemicals
Catalog number215700050
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
241.00 
Save 36.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sodium oxide (Na2O)=<0.1 %
Silicon dioxide (SiO2)=<0.04 %
Appearance (Color)White
Aluminium oxide (Al2O3)>=99.70 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatography
RUO – Research Use Only