Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Calcium phosphate, dibasic, 97%, pure, anhydrous
Thermo Scientific Chemicals

Calcium phosphate, dibasic, 97%, pure, anhydrous

CAS: 7757-93-9 | CaHO4P | 136.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 215710010
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.60
Save 18.95 (19%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.60
Save 18.95 (19%)
Each
Add to cart
Chemical Identifiers
CAS7757-93-9
IUPAC Namecalcium hydrogen phosphate
Molecular FormulaCaHO4P
InChI KeyFUFJGUQYACFECW-UHFFFAOYSA-L
SMILES[Ca++].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White
Titration Complexometric>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link