Thermo Scientific Chemicals

Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals

Catalog number: 215730500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals

Catalog number: 215730500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215730500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
688-73-3
IUPAC Name
tributylstannyl
Molecular Formula
C12H27Sn
InChI Key
PIILXFBHQILWPS-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)CCCC
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Refractive index
1.4720 to 1.4740 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text