Thermo Scientific Chemicals

Phenylmethanesulfonyl fluoride, 99%

Catalog number: 215740050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenylmethanesulfonyl fluoride, 99%

Catalog number: 215740050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215740050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
329-98-6
IUPAC Name
phenylmethanesulfonyl fluoride
Molecular Formula
C7H7FO2S
InChI Key
YBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILES
FS(=O)(=O)CC1=CC=CC=C1
Appearance (Color)
White
Infrared spectrum
Conforms
Appearance (Form)
Needles and/or agglomerates
Melting point
90°C to 94°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text