Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Phenylmethanesulfonyl fluoride, 99%
Thermo Scientific Chemicals

Phenylmethanesulfonyl fluoride, 99%

CAS: 329-98-6 | C7H7FO2S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 215740500
Price (USD)
718.00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
718.00
Each
Add to cart
Chemical Identifiers
CAS329-98-6
IUPAC Namephenylmethanesulfonyl fluoride
Molecular FormulaC7H7FO2S
InChI KeyYBYRMVIVWMBXKQ-UHFFFAOYSA-N
SMILESFS(=O)(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Needles and/or agglomerates
Melting point90°C to 94°C
Appearance (Color)White
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link