Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number 215750025
Price (USD)/ Each
438.65
Online exclusive
487.00 
Save 48.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
438.65
Online exclusive
487.00 
Save 48.35 (10%)
Add to cart
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number215750025
Price (USD)/ Each
438.65
Online exclusive
487.00 
Save 48.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-56-3
IUPAC Nametricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular FormulaO5P2
InChI KeyDLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILESO=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium (Na)=<0.005 %
Chloride (Cl)=<0.001 %
Total nitrogen (as N)=<0.002 %
Calcium (Ca)=<0.005 %
Magnesium (Mg)=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only