Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals
Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 215760500
Price (USD)/ Each
170.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
170.00
Add to cart
Bis(cyclopentadienyl)titanium dichloride, 97%, Thermo Scientific Chemicals
Catalog number215760500
Price (USD)/ Each
170.00
-
Add to cart
Chemical Identifiers
CAS1271-19-8
IUPAC Nametitanium(2+) bis(cyclopenta-2,4-dien-1-ide) dichloride
Molecular FormulaC10H10Cl2Ti
InChI KeyMKNXBRLZBFVUPV-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ti++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to bordeaux
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration Argentometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only