BS(PEG)9
BS(PEG)9
Thermo Scientific™

BS(PEG)9

BS(PEG)9 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. TheRead more
Have Questions?
Catalog NumberQuantity
21582100 mg
Catalog number 21582
Price (USD)
532.00
Each
In stock
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
532.00
Each
Add to cart
BS(PEG)9 is a bis-succinimide ester-activated PEG compound used for crosslinking between primary amines (—NH2) in proteins and other molecules. The N-hydroxysuccinimide ester (NHS) groups at either end of the PEG9 spacer react specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds.

Features of BS(PEG)9:

Reactive groups: NHS ester (both ends)
Reactive towards: amino groups (primary amines)
• NHS esters react with—NH2 groups at pH 7-9, forming stable amide bonds
• Water-soluble; compare to non-pegylated BS3 (also soluble) and DSS (water-insoluble)
• Membrane-impermeable, allowing for cell surface labeling
• Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
• Polyethylene glycol spacer arms help maintain conjugate solubility
• Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Ideal for small molecule or peptide conjugations

Alternative Names for BS(PEG)n:

• Bis-NHS-dPEG
• NHS-PEG-NHS
• BS(PEO)5
• Bis(NHS)PEO5
• BS-PEO9
• dPEG NHS ester
• Bis-succinimide ester PEG 400

Product References
Crosslinker Application Guide -- search for recent literature references for this product
Bioconjugate Techniques, Academic Press (Part No. 20036)

Related Products
BS(PEG)5 (PEGylated bis(sulfosuccinimidyl)suberate)
BS3 (bis(sulfosuccinimidyl)suberate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionBS(PEG)9
FormLiquid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)708.71
PEGylatedYes
Product LinePierce™
Quantity100 mg
Reactive MoietyNHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length35.8 Å
Water SolubleYes
Chemical ReactivityAmine-Amine
CleavableNo
Crosslinker TypeHomobifunctional
FormatStandard
Product TypeCrosslinker
SpacerLong (>30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at -20°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate)?

BS(PEG)9 (PEGylated bis(sulfosuccinimidyl)suberate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Do you recommend crosslinking reagents for viral capsids (diameter 30 nm)?

Amine-to-amine crosslinkers are a good choice for your application since amino groups are usually abundant on the surface of proteins. Our most popular amine-reactive homobifunctional crosslinker that is water-soluble is BS3 (Cat. No. A39266). This has a moderate-length (11.4 A) linker that is good for linking adjacent proteins. If you need the ability to cleave the linker with sulfhydryl reducing agents I would suggest DTSSP (Cat. No. 21578). If you think you might need an extra-long crosslinker, then BS(PEG)5 (Cat. No. 21581) or BS(PEG)9 (Cat. No. 21582) are alternatives.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection