Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals
Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 215821000
Price (USD)/ Each
65.80
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
65.80
Add to cart
Ethanesulfonyl chloride, 98+%, Thermo Scientific Chemicals
Catalog number215821000
Price (USD)/ Each
65.80
-
Add to cart
Chemical Identifiers
CAS594-44-5
IUPAC Nameethanesulfonyl chloride
Molecular FormulaC2H5ClO2S
InChI KeyFRYHCSODNHYDPU-UHFFFAOYSA-N
SMILESCCS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Form)Liquid
AppearanceMay darken on storage
Appearance (Color)Clear colorless to yellow to light brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only