Thermo Scientific Chemicals

Ethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 215835000
500 g, Each
Thermo Scientific Chemicals

Ethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 215835000
500 g, Each
Quantity
Catalog number: 215835000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
97-63-2
IUPAC Name
ethyl 2-methylprop-2-enoate
Molecular Formula
C6H10O2
InChI Key
SUPCQIBBMFXVTL-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)=C
Appearance (Form)
Liquid
GC
>=98.5 %
Stabilizer
10 to 120 ppm MEHQ
Appearance (Color)
Clear colorless
Refractive index
1.4120 to 1.4160 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text