Thermo Scientific Chemicals

Butyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals

Catalog number: 215860025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Butyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals

Catalog number: 215860025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 215860025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-32-2
IUPAC Name
butyl prop-2-enoate
Molecular Formula
C7H12O2
InChI Key
CQEYYJKEWSMYFG-UHFFFAOYSA-N
SMILES
CCCCOC(=O)C=C
Color scale
=<10 APHA
GC
>=99.0 %
Acidity
=<0.01 % (as Acrylic acid)
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text