Thermo Scientific Chemicals

Chlorotriethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 215940250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Chlorotriethylsilane, 99%, Thermo Scientific Chemicals

Catalog number: 215940250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 215940250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
994-30-9
IUPAC Name
chlorotriethylsilane
Molecular Formula
C6H15ClSi
InChI Key
DCFKHNIGBAHNSS-UHFFFAOYSA-N
SMILES
CC[Si](Cl)(CC)CC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=98.5 %
Refractive index
1.4300 to 1.4320 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text