Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals
Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 215960050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namesodium bis(trimethylsilyl)azanide
Molecular FormulaC6H18NNaSi2
InChI KeyWRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES[Na+].C[Si](C)(C)[N-][Si](C)(C)C
Molecular Weight (g/mol)183.38
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO495.0 to 105.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only