Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals

Catalog number: 215960250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals

Catalog number: 215960250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 215960250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1070-89-9
IUPAC Name
sodium bis(trimethylsilyl)azanide
Molecular Formula
C6H18NNaSi2
InChI Key
WRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES
[Na+].C[Si](C)(C)[N-][Si](C)(C)C
Appearance (Color)
White to light yellow to light beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
95.0 to 105.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text