Thermo Scientific Chemicals

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: 216110250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog number: 216110250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 216110250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38078-09-0
IUPAC Name
diethyl(trifluoro-λ⁴-sulfanyl)amine
Molecular Formula
C4H10F3NS
InChI Key
CSJLBAMHHLJAAS-UHFFFAOYSA-N
SMILES
CCN(CC)S(F)(F)F
Appearance (Color)
Clear yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
NMR (1H-NMR)
>=94 %
Refractive index
1.4100 to 1.4160 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text