Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals
Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Plastic bottle
Catalog number 216111000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS38078-09-0
IUPAC Namediethyl(trifluoro-λ⁴-sulfanyl)amine
Molecular FormulaC4H10F3NS
InChI KeyCSJLBAMHHLJAAS-UHFFFAOYSA-N
SMILESCCN(CC)S(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
NMR (1H-NMR)>=94 %
Refractive index1.4100 to 1.4160 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only