Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SPLVLQSDRN VTVNARNHMG QLTGQLTIGA DAVEAQCKRF EVRASEDGRV LFSADEDEIT IGAEKLKVTG TEGAVFGHSV ETPHIRAEPS QDLRLESPTR SLIMEAPRGV QVSAAAGDFK ATCRKELHLQ STEGEIFLNA ETIKLGNLPT GSFSSSSPSS SSSRQTVYEL CVCPNGKLYL SPAGVGSTCQ SSSNICLWS
The zeta-sarcoglycan gene measures over 465 kb and localizes to 8p22. This protein is part of the sarcoglycan complex, a group of 6 proteins. The sarcoglycans are all N-glycosylated transmembrane proteins with a short intra-cellular domain, a single transmembrane region and a large extra-cellular domain containing a carboxyl-terminal cluster with several conserved cysteine residues. The sarcoglycan complex is part of the dystrophin-associated glycoprotein complex (DGC), which bridges the inner cytoskeleton and the extra-cellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: sarcoglycan, zeta; Zeta-sarcoglycan; Zeta-SG; ZSG1
Gene Aliases: C230085N17Rik; SGCZ; ZSG1
UniProt ID: (Human) Q96LD1, (Mouse) Q8BX51
Entrez Gene ID: (Human) 137868, (Mouse) 244431, (Rat) 364605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support