tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 216221000
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
Add to cart
tert-Butyl nitrite, 90%, pure, Thermo Scientific Chemicals
Catalog number216221000
Price (USD)/ Each
76.65
Online exclusive
85.20 
Save 8.55 (10%)
-
Add to cart
Chemical Identifiers
CAS540-80-7
IUPAC Nametert-butyl nitrite
Molecular FormulaC4H9NO2
InChI KeyIOGXOCVLYRDXLW-UHFFFAOYSA-N
SMILESCC(C)(C)ON=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=88 %
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.3670 to 1.3700 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only