Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, Thermo Scientific Chemicals
Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 216270051
Price (USD)/ Each
516.00
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
516.00
Add to cart
Methyl-5-norbornene-2,3-dicarboxylic anhydride, 95%, mixture of isomers, Thermo Scientific Chemicals
Catalog number216270051
Price (USD)/ Each
516.00
-
Add to cart
Chemical Identifiers
CAS25134-21-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.5060 to 1.5080 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only