Thermo Scientific Chemicals

Hexamethylcyclotrisiloxane, 98%, Thermo Scientific Chemicals

Catalog number: 216460250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Hexamethylcyclotrisiloxane, 98%, Thermo Scientific Chemicals

Catalog number: 216460250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 216460250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
541-05-9
IUPAC Name
hexamethyl-1,3,5,2,4,6-trioxatrisilinane
Molecular Formula
C6H18O3Si3
InChI Key
HTDJPCNNEPUOOQ-UHFFFAOYSA-N
SMILES
C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O1
Appearance (Color)
Colorless to white
Appearance (Form)
Crystalline chunks
Infrared spectrum
Conforms
Melting point
63°C to 67°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text