Thermo Scientific Chemicals

Octyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: 216501000
100 g, Each
Thermo Scientific Chemicals

Octyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog number: 216501000
100 g, Each
Quantity
Catalog number: 216501000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5283-66-9
IUPAC Name
trichloro(octyl)silane
Molecular Formula
C8H17Cl3Si
InChI Key
RCHUVCPBWWSUMC-UHFFFAOYSA-N
SMILES
CCCCCCCC[Si](Cl)(Cl)Cl
Appearance (Color)
Clear colorless to light yellow to light brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.4470 to 1.4490 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text