Thermo Scientific Chemicals

1,1,3,3-Tetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 216515000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,1,3,3-Tetramethyldisiloxane, 97%, Thermo Scientific Chemicals

Catalog number: 216515000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 216515000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3277-26-7
Molecular Formula
C4H12OSi2
InChI Key
KWEKXPWNFQBJAY-UHFFFAOYSA-N
SMILES
C[Si](C)O[Si](C)C
Molecular Weight (g/mol)
132.31
Refractive index
1.3690 to 1.3710 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text