Thermo Scientific Chemicals

Vinyltrimethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: 216521000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Vinyltrimethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: 216521000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 216521000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2768-02-7
IUPAC Name
ethenyltrimethoxysilane
Molecular Formula
C5H12O3Si
InChI Key
NKSJNEHGWDZZQF-UHFFFAOYSA-N
SMILES
CO[Si](OC)(OC)C=C
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=97.5 %
Refractive index
1.3915 to 1.3935 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text