Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 216620010
Price (USD)/ Each
49.65
Online exclusive
55.00 
Save 5.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
55.00 
Save 5.35 (10%)
Add to cart
Butyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals
Catalog number216620010
Price (USD)/ Each
49.65
Online exclusive
55.00 
Save 5.35 (10%)
-
Add to cart
Chemical Identifiers
CAS97-88-1
IUPAC Namebutyl 2-methylprop-2-enoate
Molecular FormulaC8H14O2
InChI KeySOGAXMICEFXMKE-UHFFFAOYSA-N
SMILESCCCCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=98.5 %
Refractive index1.4225 to 1.4245 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only