Thermo Scientific Chemicals

Folic acid, 97%, pure

Catalog number: 216630500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Folic acid, 97%, pure

Catalog number: 216630500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 216630500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59-30-3
IUPAC Name
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Molecular Formula
C19H19N7O6
InChI Key
OVBPIULPVIDEAO-LBPRGKRZSA-N
SMILES
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
Appearance (Color)
Yellow to orange
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Sulfated ash
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text