Chemical Identifiers
CAS59-30-3
IUPAC Name(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Molecular FormulaC19H19N7O6
InChI KeyOVBPIULPVIDEAO-LBPRGKRZSA-N
SMILESNC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
View more