Thermo Scientific™

DMP (dimethyl pimelimidate)

Catalog number: 21666
Have Questions?
Thermo Scientific™

DMP (dimethyl pimelimidate)

Catalog number: 21666
Have Questions?
Catalog Number
21666
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2166650 mg
-
216671 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.

Features of dimethyl pimelimidate:

Reactive groups: imidoester (both ends)
Reactive towards: amino groups (primary amines)
• Mid-length reagent of three similar imidoester crosslinkers, which include DMA, DMP and DMS
• Water-soluble
• Imidoesters react with amines at alkaline pH values (pH 8-10) to form amidine bond
• Amidine bond retains net charge character of protein primary amine to which it reacts
• Reversible at high pH values
• Tool for study of quarternary structure of proteins

Product References
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
DMA (dimethyl adipimidate)
DMS (dimethyl suberimidate)
DTBP (Wang and Richard's Reagent)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Amine
Cleavable
No
Description
DMP
Molecular Weight (g/mol)
259.17
PEGylated
No
Spacer Arm Length
9.2 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homobifunctional
Reactive Moiety
Imidoester
Spacer
Short (<10 Å)
Form
Powder
Quantity
50 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text