Thermo Scientific Chemicals

Aluminum fluoride trihydrate, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 216825000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Aluminum fluoride trihydrate, 97%, extra pure, Thermo Scientific Chemicals

Catalog number: 216825000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 216825000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15098-87-0
IUPAC Name
aluminium(3+) trihydrate trifluoride
Molecular Formula
AlF3H6O3
InChI Key
BKLZIAYVINRQEJ-UHFFFAOYSA-K
SMILES
O.O.O.[F-].[F-].[F-].[Al+3]
Appearance (Color)
White
Appearance (Form)
Powder
Assay
>=96.0 %
Iron (Fe)
=<100 ppm
Sulfate (SO4)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text