Thermo Scientific Chemicals

Ethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 217042500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethanolamine hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 217042500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 217042500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2002-24-6
IUPAC Name
hydrogen 2-aminoethan-1-ol chloride
Molecular Formula
C2H8ClNO
InChI Key
PMUNIMVZCACZBB-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NCCO
Infrared spectrum
Conforms
Appearance (Form)
Crystals and/or agglomerates
Appearance (Color)
White to light beige
Titration Argentometric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text