Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 217095000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 217095000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 217095000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-77-1
IUPAC Name
iron(3+) hexahydrate trichloride
Molecular Formula
Cl3FeH12O6
InChI Key
NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Appearance (Form)
Adhering crystalline powder and/or lumps
Phosphorous compounds (as PO4)
=<100 ppm
Copper (Cu)
=<30 ppm
Zinc (Zn)
=<30 ppm
Titration Iodometric
99.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text