Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 217100010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH7.5 to 9.0 (5% soln. at 20°C)
Chloride (Cl)=<50 ppm
Appearance (Form)Granular powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only