Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 217105000
Price (USD)/ Each
46.65
Online exclusive
54.20 
Save 7.55 (14%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
46.65
Online exclusive
54.20 
Save 7.55 (14%)
Add to cart
Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number217105000
Price (USD)/ Each
46.65
Online exclusive
54.20 
Save 7.55 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
Molecular Weight (g/mol)98.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH7.5 to 9.0 (5% soln. at 20°C)
Chloride (Cl)=<50 ppm
Appearance (Form)Granular powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only