Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 217110010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 217110010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 217110010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6131-90-4
IUPAC Name
sodium acetate trihydrate
Molecular Formula
C2H9NaO5
InChI Key
AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CC([O-])=O
Aluminium (Al)
=<5 ppm
Appearance (Color)
White
Heavy metals (as Pb)
=<5 ppm
Phosphate (PO4)
=<5 ppm
Iron (Fe)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text