Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Acetic acid, sodium salt trihydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99.5%, for analysis

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 217115000
Price (USD)
51.60
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
51.60
Each
Add to cart
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Aluminium (Al)=<5 ppm
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Phosphate (PO4)=<5 ppm
Iron (Fe)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link