Thermo Scientific Chemicals

Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 217120010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 217120010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 217120010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-55-8
IUPAC Name
sodium hydrogen carbonate
Molecular Formula
CHNaO3
InChI Key
UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES
[Na+].OC([O-])=O
Titration with HCl
>=99.4 %
Calcium (Ca)
=<100 ppm
Potassium (K)
=<50 ppm
Magnesium (Mg)
=<50 ppm
Zinc (Zn)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text